Learn More
Chemglass Life Sciences Pack, Blue Bonded Closure and Vial, Glass, 2.0mL, 9mm Thread, Clear with Grad Spot, 12 x 32mm, PTFE/Red Rubber with Slit
SDP
Catalog No. 501532656
Encompass_Preferred
Pack, Blue Bonded Closure and Vial, Glass, 2.0mL, 9mm Thread, Clear with Grad Spot, 12 x 32mm, PTFE/Red Rubber with Slit. These convenience packs come with 100 closures and vials packed seperately. Closures are not attached.
Catalog No. 50-153-2656 Supplier Chemglass Life Sciences Supplier No. CV53261233
Pack, Blue Bonded Closure and Vial, Glass, 2.0mL, 9mm Thread, Clear with Grad Spot, 12 x 32mm, PTFE/Red Rubber with Slit. These convenience packs come with 100 closures and vials packed seperately. Closures are not attached.