Learn More
Waters Corp Waters Tube Collection Rack, 5 mL Tube, 12-Position, Andrew+ Compatible
Catalog No. 502269418 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This consumable tube collection rack is designed to hold 5 mL tubes for various laboratory applications. It provides a secure and organized solution for sample handling and is compatible with automated systems for increased efficiency.
  • Designed to hold 12x 5 mL tubes.
  • Compatible with the Extraction+ Device.
  • Compatible with the Collection Rack Domino.
  • Can be moved by the Andrew+ robot when utilizing the Microplate Gripper tool.
This item is not returnable. View return policy
Catalog No. 50-226-9418 Supplier Waters Corp Supplier No. 186010534
This consumable tube collection rack is designed to hold 5 mL tubes for various laboratory applications. It provides a secure and organized solution for sample handling and is compatible with automated systems for increased efficiency.
  • Designed to hold 12x 5 mL tubes.
  • Compatible with the Extraction+ Device.
  • Compatible with the Collection Rack Domino.
  • Can be moved by the Andrew+ robot when utilizing the Microplate Gripper tool.