Learn More
STRATASYS INC PACK OF 1 RGD531S, POLYJET 531S REINFORCER, 4 KG
Supplier:  STRATASYS INC OBJ18078
Encompass
PolyJet™ 531S Reinforcer (last name: RGD531, Digital ABS Plus) is designed to simulate standard ABS plastics, combining high-temperature resistance with excellent toughness. Ideal for parts requiring PolyJet™ technology’s highest impact resistance and shock absorption, PolyJet™ 531S Reinforcer significantly enhances mechanical performance for design verification and functional testing. It delivers improved impact strength along with high thermal resistance, durability, and a superior surface finish. The material is well-suited for rapid prototyping of snap-fit parts used in high or low temperatures, functional designs requiring multi-material flexibility, as well as molds, manufacturing tools, electrical components, and more.The new formulation of PolyJet™ 531S Reinforcer is a TPO-free version. The updated cartridges carry the same RFID as the previous RGD531 material, and when inserted into the cabinet, the printer will identify them
Catalog No. NC3887962