Learn More
Waters Corp Waters Sealing Caps, 1 cc, 24 per pack
Catalog No. 502269419 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The 1 cc Sealing Caps are compatible with the 1 cc Cartridge Adaptor for Extraction+. They are used to block/seal any unused positions during extraction runs so as to optimize and homogenize pressure flow.
  • Compatible with the 1 cc Cartridge Adaptor for Extraction+
  • Used to block/seal any unused positions during extraction runs
  • Optimizes and homogenizes pressure flow
  • 24 caps per pack
  • 1 cc size
  • One pack included
Catalog No. 50-226-9419 Supplier Waters Corp Supplier No. 186010535
The 1 cc Sealing Caps are compatible with the 1 cc Cartridge Adaptor for Extraction+. They are used to block/seal any unused positions during extraction runs so as to optimize and homogenize pressure flow.
  • Compatible with the 1 cc Cartridge Adaptor for Extraction+
  • Used to block/seal any unused positions during extraction runs
  • Optimizes and homogenizes pressure flow
  • 24 caps per pack
  • 1 cc size
  • One pack included