Learn More
Waters Corp Waters Sealing Caps, 3cc, 24 pk
Supplier: Waters Corp 186010536
The 3cc Sealing Caps are designed for use with the 3cc Cartridge Adaptor for Extraction+. They effectively block and seal any unused positions during extraction runs.
- Compatible with 3cc cartridge adaptor for extraction+.
- Used to block/seal unused positions during extraction runs.
- Optimizes and homogenizes pressure flow during extraction.