Learn More
Lonza Walkersville Eppendorf 2–200 µL Biopur Pipette Tips
Catalog No. NC9041347
Encompass
Eppendorf 2–200 µL Biopur Pipette Tips are to be used when testing with any of our Endotoxin Detection Test Kits. The tips are certified to contain <0.001 EU/ml endotoxin. The new design of the tips allows more flexibility with different pipettors.Benefits:Certified to contain less than 0.001 EU/mlContent:5 trays/package: 96 tips/tray
Catalog No. NC9041347 Supplier Lonza Walkersville Supplier No. 25415
Eppendorf 2–200 µL Biopur Pipette Tips are to be used when testing with any of our Endotoxin Detection Test Kits. The tips are certified to contain <0.001 EU/ml endotoxin. The new design of the tips allows more flexibility with different pipettors.Benefits:Certified to contain less than 0.001 EU/mlContent:5 trays/package: 96 tips/tray