Learn More
Med Vet International Attest Rapid 5 Steam-Plus Sterilization Biological Indicator Pack Steam 2 inch, 41382, 16/Box
Catalog No. 502829366
Encompass_Preferred
3M Attest rapid 5 steam-plus test packs are designed to monitor 270° F vacuum-assisted steam sterilization cycles 4 minutes in length and 250° F gravity-displacement steam sterilization cycles 30 minutes in length Brown cap Includes 16 test packs and 16 controls Consists of 1292 BI + Comply™ (SteriGage™) Steam Chemical Integrator
Catalog No. 50-282-9366 Supplier Med Vet International Supplier No. 566792BX
Add to Cart
Add to Cart
3M Attest rapid 5 steam-plus test packs are designed to monitor 270° F vacuum-assisted steam sterilization cycles 4 minutes in length and 250° F gravity-displacement steam sterilization cycles 30 minutes in length Brown cap Includes 16 test packs and 16 controls Consists of 1292 BI + Comply™ (SteriGage™) Steam Chemical Integrator