Learn More
Med Vet International McKesson Iodoform Wound Packing Strip, ½ Inch x 5 Yard, 61-59245, Each
Catalog No. 503043186
Encompass_Preferred
Wound Packing Strip McKesson Antiseptic Cotton Iodoform Medium 1/2 Inch X 5 Yard 1 Count Sterile Easy dispensing packaging (patent pending) helps facilitate caregiver convenience Made of 100% cotton, ravel-resistant gauze designed to resist linting or fraying Conveniently designed for secure storage and extended shelf life Sterile until opened ½ inch x 5 yards
Catalog No. 50-304-3186 Supplier Med Vet International Supplier No. 470441BT
Wound Packing Strip McKesson Antiseptic Cotton Iodoform Medium 1/2 Inch X 5 Yard 1 Count Sterile Easy dispensing packaging (patent pending) helps facilitate caregiver convenience Made of 100% cotton, ravel-resistant gauze designed to resist linting or fraying Conveniently designed for secure storage and extended shelf life Sterile until opened ½ inch x 5 yards