Learn More
HAUG QUALITY EQUIPMENT PACK-VAC LEAK DET PVCA201413
Catalog No. NC2309718
Encompass
  • Compressed air-powered package leak detector
  • Conducts precise assessments of package seal integrity
  • Utilizes venturi vacuum generator
  • Bubble emission test method for visual leak identification
  • Test duration is less than 30 seconds
  • Tank dimensions: 20 in x 14 in x 13 in
  • 360° viewing through a clear tank
  • Requires 60-80 PSI, 2-6 CFM of standard dry plant air
  • Suitable for abusive production environments
  • Applicable for leak detection, seal integrity testing, and altitude simulation
  • Meets ASTM standards D3078, D6653, D4169, D4991
Catalog No. NC2309718 Supplier HAUG QUALITY EQUIPMENT Supplier No. PVCA201413
Add to Cart
Add to Cart
Request a Quote
  • Compressed air-powered package leak detector
  • Conducts precise assessments of package seal integrity
  • Utilizes venturi vacuum generator
  • Bubble emission test method for visual leak identification
  • Test duration is less than 30 seconds
  • Tank dimensions: 20 in x 14 in x 13 in
  • 360° viewing through a clear tank
  • Requires 60-80 PSI, 2-6 CFM of standard dry plant air
  • Suitable for abusive production environments
  • Applicable for leak detection, seal integrity testing, and altitude simulation
  • Meets ASTM standards D3078, D6653, D4169, D4991