Learn More
BURKLE INC Packaging bucket, PP white, 1.32 gal. (5l), with closure
Catalog No. 5000025843
Encompass_Preferred
Packaging bucket for transport, packaging and storing of various materials. Suitable for the food or building material industries and non-aggressive chemicals in environmental protection, hospitals and the household. With strong handle. Quality closure on the bucket for product safety.
Catalog No. 50-000-25843 Supplier BURKLE INC Supplier No. 23270050
Packaging bucket for transport, packaging and storing of various materials. Suitable for the food or building material industries and non-aggressive chemicals in environmental protection, hospitals and the household. With strong handle. Quality closure on the bucket for product safety.