Learn More
Perkin Elmer US LLC Packed Column Adapters - 0.25 in, Pkg. 2
Catalog No. 502117203
Encompass_Preferred
This 1/8 inch to 1/4 inch detector reciever adapter is for use with 1/4 inch packed columns and fits both the detector and injector outlet (inside of oven). Package of two (injector/detector). Includes the following pieces: 2 Adapter fittings, 1/4 inch column to 1/8 inch fitting (00081257) 2 Stainless Steel Hex Nuts, 1/8 inch (09903453) 2 Stainless Steel Front Ferrules, 1/8 inch (09903454) 2 Stainless Steel Back Ferrules, 1/8 inch (09903455)
Catalog No. 50-211-7203 Supplier Perkin Elmer US LLC Supplier No. 00080100
This 1/8 inch to 1/4 inch detector reciever adapter is for use with 1/4 inch packed columns and fits both the detector and injector outlet (inside of oven). Package of two (injector/detector). Includes the following pieces: 2 Adapter fittings, 1/4 inch column to 1/8 inch fitting (00081257) 2 Stainless Steel Hex Nuts, 1/8 inch (09903453) 2 Stainless Steel Front Ferrules, 1/8 inch (09903454) 2 Stainless Steel Back Ferrules, 1/8 inch (09903455)