Learn More
Perkin Elmer US LLC Packed Column Injector Kit with PPC, 120 V
Catalog No. 502113740
Encompass_Preferred
The GC must be PPC (Programmable Pneumatic Control) ready. If not, a PPC upgrade kit (N6120146) is required.
Catalog No. 50-211-3740 Supplier Perkin Elmer US LLC Supplier No. N6120136
Add to Cart
Add to Cart
Request a Quote
The GC must be PPC (Programmable Pneumatic Control) ready. If not, a PPC upgrade kit (N6120146) is required.