Learn More
Perkin Elmer US LLC Packed Column Injector Starter Kit
Catalog No. 502111615
Encompass_Preferred
Includes the following pieces: Silanized Glass Wool, 2 oz (03300905) Packed Injector Liner (N6101048) Packed Injector Needle Guide (N6101050) Liner Removal Tool (N6100102) Capillary Injector Packing Tool (N610T100)
Catalog No. 50-211-1615 Supplier Perkin Elmer US LLC Supplier No. N6120100
Includes the following pieces: Silanized Glass Wool, 2 oz (03300905) Packed Injector Liner (N6101048) Packed Injector Needle Guide (N6101050) Liner Removal Tool (N6100102) Capillary Injector Packing Tool (N610T100)