Learn More
Stoelting™ Packer Mosquito Hemostatic Forceps
SDP
Catalog No. 10001325 Shop All Stoelting Co Products
Change view
Click to view available options
Model:
Curved, Flat
Straight, Flat
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model
10-001-325 Curved, Flat
10-001-324 Straight, Flat
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 10-001-325 Supplier Stoelting™ Supplier No. 5211005
Add to Cart
Add to Cart
Stoelting™ Packer Mosquito Hemostatic Forceps are used in many surgical procedures usually to control bleeding or the flow of liquids in tubing.
  • Scissor-like ring handles just like scissors
  • Ratcheted handles can be locked in multiple positions in order to maintain variable levels of constant pressure
  • Delicate hemostats have thin jaws with fine serrations
Specifications
For Use With (Application) Neuroscience
Length (English) 5 in.
Model Curved, Flat