Learn More
Microsolv Technology Corporation Packing Adapter, PEEK, 7.5mm ID, Column Packing Hardware, for use with the All-PEEK Column Packing System. 1 EA.
Catalog No. 502271573
Encompass_Preferred
This Packing Adapter is made for 7.5mm ID PEEK Columns with 5/8"-18 screw threads and is designed for packing only All-PEEK HPLC Column Hardware and not recommended for use with stainless steel packing systems. This adapter is made from Victrex Brand PEEK which ensures a biocompatibility from the beginning of the packing and column process.
This item is not returnable. View return policy
Catalog No. 50-227-1573 Supplier Microsolv Technology Corporation Supplier No. 49333PA3
This Packing Adapter is made for 7.5mm ID PEEK Columns with 5/8"-18 screw threads and is designed for packing only All-PEEK HPLC Column Hardware and not recommended for use with stainless steel packing systems. This adapter is made from Victrex Brand PEEK which ensures a biocompatibility from the beginning of the packing and column process.