Learn More
Med Vet International McKesson Nonimpregnated Wound Packing Strip, 1 Inch x 5 Yard, 61-59320, 12/Case
Catalog No. 503045443
Encompass_Preferred
Wound Packing Strip McKesson Plain Cotton Non-impregnated Large 1 Inch X 5 Yard 1 Count Sterile Easy dispensing packaging (patent pending) helps facilitate caregiver convenience Made of 100% cotton, ravel-resistant gauze designed to resist linting or fraying Conveniently designed for secure storage and extended shelf life 1 inch x 5 yards
Catalog No. 50-304-5443 Supplier Med Vet International Supplier No. 470438CS
 Restricted Item
Add to Cart
Add to Cart
Wound Packing Strip McKesson Plain Cotton Non-impregnated Large 1 Inch X 5 Yard 1 Count Sterile Easy dispensing packaging (patent pending) helps facilitate caregiver convenience Made of 100% cotton, ravel-resistant gauze designed to resist linting or fraying Conveniently designed for secure storage and extended shelf life 1 inch x 5 yards