Learn More
Waters Corp Waters Corporation Packing Ring, Nebulizer, Red Silicone, N-type Fluoropolymer Coated
Catalog No. 50791836 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This packing ring is designed for nebulizers, providing a reliable seal. It is made from red silicone with an N-type Fluoropolymer coating, ensuring durability and chemical resistance.
  • Made of red silicone with N-type Fluoropolymer coating.
  • Designed as a spare part.
  • Provides a reliable seal for nebulizers.
Catalog No. 50-791-836 Supplier Waters Corp Supplier No. 425000326
This packing ring is designed for nebulizers, providing a reliable seal. It is made from red silicone with an N-type Fluoropolymer coating, ensuring durability and chemical resistance.
  • Made of red silicone with N-type Fluoropolymer coating.
  • Designed as a spare part.
  • Provides a reliable seal for nebulizers.