Learn More
Microsolv Technology Corporation Packing Seal, PEEK, 7.5mm ID, Column Packing Hardware, for use with All-PEEK Column Packing System. 1 EA.
Catalog No. 502271575
Encompass_Preferred
This Packing Seal is made for use with the Packing Adapter and 7.5mm ID PEEK Columns. Used for packing All-PEEK HPLC Column Hardware and not recommended for use with stainless steel packing systems. This Seal is made from Victrex Brand PEEK which ensures a biocompatibility from the beginning of the packing and column process.
This item is not returnable. View return policy
Catalog No. 50-227-1575 Supplier Microsolv Technology Corporation Supplier No. 49333PS
This Packing Seal is made for use with the Packing Adapter and 7.5mm ID PEEK Columns. Used for packing All-PEEK HPLC Column Hardware and not recommended for use with stainless steel packing systems. This Seal is made from Victrex Brand PEEK which ensures a biocompatibility from the beginning of the packing and column process.