Learn More
Wilmad Labglass PACKING SOLID GLASS BEADS 4MM
Supplier:  Wilmad Labglass LG6750102
Encompass
Distillation Column Packing, Solid Glass Beads. Approximate Diameter: 4mm; Approximate Number Per Pound: 5,500. Manufactured from borosilicate glass. Sold in one pound packages.
Catalog No. 50-800-456