Learn More
Med Vet International Dynarex Packing Strip, Iodoform, Sterile, 2" X 5 Yards, 12/Case
Catalog No. 502829929
Encompass_Preferred
Sterile iodoform packing strips are ideal for sterile drainage of open/infected wounds. Ideal for use as nasal packing, drainage wicks, or for packing wounds Specially woven gauze will not ravel, lint or fray Impregnated with an iodoform solution 100% Cotton
Catalog No. 50-282-9929 Supplier Med Vet International Supplier No. 3414
Sterile iodoform packing strips are ideal for sterile drainage of open/infected wounds. Ideal for use as nasal packing, drainage wicks, or for packing wounds Specially woven gauze will not ravel, lint or fray Impregnated with an iodoform solution 100% Cotton