Learn More
Med Vet International Dynarex Packing Strip Plain, Sterile, 2" X 5 Yards, 12/Case
Catalog No. 502829916
Encompass_Preferred
Sterile plain packing strips ideal for sterile drainage of open/infected wounds. Ideal for use as nasal packing, drainage wicks, or for packing wounds Specially woven gauze will not ravel, lint or fray 100% Cotton
Catalog No. 50-282-9916 Supplier Med Vet International Supplier No. 3404
 Restricted Item
Add to Cart
Add to Cart
Sterile plain packing strips ideal for sterile drainage of open/infected wounds. Ideal for use as nasal packing, drainage wicks, or for packing wounds Specially woven gauze will not ravel, lint or fray 100% Cotton