Learn More
Med Vet International McKesson Iodoform Wound Packing Strip, 1 Inch x 5 Yard, 61-59345, 12/Case
Catalog No. 503044265
Encompass_Preferred
Wound Packing Strip McKesson Antiseptic Cotton Iodoform Large 1 Inch X 5 Yard 1 Count Sterile Easy dispensing packaging (patent pending) helps facilitate caregiver convenience Made of 100% cotton, ravel-resistant gauze designed to resist linting or fraying Conveniently designed for secure storage and extended shelf life Sterile until opened 1 inch x 5 yards
Catalog No. 50-304-4265 Supplier Med Vet International Supplier No. 470442CS
Wound Packing Strip McKesson Antiseptic Cotton Iodoform Large 1 Inch X 5 Yard 1 Count Sterile Easy dispensing packaging (patent pending) helps facilitate caregiver convenience Made of 100% cotton, ravel-resistant gauze designed to resist linting or fraying Conveniently designed for secure storage and extended shelf life Sterile until opened 1 inch x 5 yards