Learn More
Med Vet International McKesson Nonimpregnated Wound Packing Strip, ¼ Inch x 5 Yard, 61-59120, Each
Catalog No. 503044126
Encompass_Preferred
Wound Packing Strip McKesson Plain Cotton Non-impregnated Small 1/4 Inch X 5 Yard 1 Count Sterile Easy dispensing packaging (patent pending) helps facilitate caregiver convenience Made of 100% cotton, ravel-resistant gauze designed to resist linting or fraying Conveniently designed for secure storage and extended shelf life ¼ inch x 5 yards
Catalog No. 50-304-4126 Supplier Med Vet International Supplier No. 470436BT
Wound Packing Strip McKesson Plain Cotton Non-impregnated Small 1/4 Inch X 5 Yard 1 Count Sterile Easy dispensing packaging (patent pending) helps facilitate caregiver convenience Made of 100% cotton, ravel-resistant gauze designed to resist linting or fraying Conveniently designed for secure storage and extended shelf life ¼ inch x 5 yards