Learn More
Medchemexpress LLC HY-B0015 100mg Medchemexpress, Paclitaxel CAS:33069-62-4 Purity:>98%
SDP
Catalog No. 502029382
Encompass_Preferred
Medchemexpress, HY-B0015 100mg Paclitaxel CAS:33069-62-4 Paclitaxel (Taxol) is a naturally occurring antineoplastic agent and stabilizes tubulin polymerization. Paclitaxel can cause both mitotic arrest and apoptotic cell death. Paclitaxel also induces autophagy[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9382 Supplier Medchemexpress LLC Supplier No. HYB0015100MG
Medchemexpress, HY-B0015 100mg Paclitaxel CAS:33069-62-4 Paclitaxel (Taxol) is a naturally occurring antineoplastic agent and stabilizes tubulin polymerization. Paclitaxel can cause both mitotic arrest and apoptotic cell death. Paclitaxel also induces autophagy[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.