Learn More
Selleck Chemical LLC Paclitaxel 10mM/1mL 33069-62-4 NSC 125973, PTX, Taxol, Onxal
SDP
Catalog No. NC0634287
Encompass_Preferred
Paclitaxel (NSC 125973, PTX, Taxol, Onxal) is a microtubule polymer stabilizer with IC50 of 0.1 pM in human endothelial cells. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC0634287 Supplier Selleck Chemical LLC Supplier No. S115010MM/1ML
Paclitaxel (NSC 125973, PTX, Taxol, Onxal) is a microtubule polymer stabilizer with IC50 of 0.1 pM in human endothelial cells. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.