Learn More
Medchemexpress LLC Paclitaxel-2′-succinate NHS ester | 245110-80-9 | 98.8% | 1051.05 | 1 MG
SDP
Catalog No. 5000392134
Encompass_Preferred
Paclitaxel-2′-succinate NHS ester is a derivative of paclitaxel that features a succinic acid linker, with the carboxyl group activated by an NHS ester. The NHS ester group demonstrates high reactivity with amino or hydroxyl groups.
  • Suitable for conjugation with various molecules, including peptides, proteins, antibodies, enzymes, or polymers.
  • Intended for use in the development of nanomedicines.
  • Used in cancer therapy research.
Catalog No. 50-003-92134 Supplier Medchemexpress LLC Supplier No. HY1641651MG
Paclitaxel-2′-succinate NHS ester is a derivative of paclitaxel that features a succinic acid linker, with the carboxyl group activated by an NHS ester. The NHS ester group demonstrates high reactivity with amino or hydroxyl groups.
  • Suitable for conjugation with various molecules, including peptides, proteins, antibodies, enzymes, or polymers.
  • Intended for use in the development of nanomedicines.
  • Used in cancer therapy research.