Learn More
Lkt Laboratories Paclitaxel, from Taxus yunnanensis, ≥98%, 25MG
SDP
Supplier:  Lkt Laboratories P009225MG
Encompass_Preferred
Diterpene found in Taxus yunnanensis; microtubule depolymerization inhibitor. CAS: 33069-62-4. Additional sizes available upon request. Ships from Saint Paul, MN.
Catalog No. NC9747088