Learn More
Medchemexpress LLC Paclitaxel | 33069-62-4 | 99.9% | C₄₇H₅₁NO₁₄ | 100 MG
SDP
Catalog No. 5000433415
Encompass_Preferred
Paclitaxel is an analytical standard and a naturally occurring antineoplastic agent. It is intended for research and analytical applications, stabilizing tubulin polymerization, causing mitotic arrest, apoptotic cell death, and inducing autophagy. This compound serves as a reference standard for assays and is utilized in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS.
  • Analytical standard for paclitaxel
  • Naturally occurring antineoplastic agent
  • Stabilizes tubulin polymerization
  • Causes mitotic arrest
  • Induces apoptotic cell death
  • Induces autophagy
  • Reference standard for assays
  • Used in HPLC, GC, and MS experiments
Catalog No. 50-004-33415 Supplier Medchemexpress LLC Supplier No. HYB0015R100MG
Paclitaxel is an analytical standard and a naturally occurring antineoplastic agent. It is intended for research and analytical applications, stabilizing tubulin polymerization, causing mitotic arrest, apoptotic cell death, and inducing autophagy. This compound serves as a reference standard for assays and is utilized in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS.
  • Analytical standard for paclitaxel
  • Naturally occurring antineoplastic agent
  • Stabilizes tubulin polymerization
  • Causes mitotic arrest
  • Induces apoptotic cell death
  • Induces autophagy
  • Reference standard for assays
  • Used in HPLC, GC, and MS experiments