Learn More
Medchemexpress LLC Paclitaxel | 33069-62-4 | 99.97% | C47H51NO14 | 5 G
SDP
Catalog No. 5000433414
Encompass_Preferred
Paclitaxel is a naturally occurring antineoplastic agent that stabilizes tubulin polymerization. It can cause both mitotic arrest and apoptotic cell death, and it also induces autophagy.
  • Naturally occurring antineoplastic agent.
  • Stabilizes tubulin polymerization.
  • Causes mitotic arrest.
  • Induces apoptotic cell death.
  • Induces autophagy.
Catalog No. 50-004-33414 Supplier Medchemexpress LLC Supplier No. HYB00155G
Paclitaxel is a naturally occurring antineoplastic agent that stabilizes tubulin polymerization. It can cause both mitotic arrest and apoptotic cell death, and it also induces autophagy.
  • Naturally occurring antineoplastic agent.
  • Stabilizes tubulin polymerization.
  • Causes mitotic arrest.
  • Induces apoptotic cell death.
  • Induces autophagy.