Learn More
Medchemexpress LLC Paclitaxel | 33069-62-4 | C47H51NO14 | 1 G
SDP
Catalog No. 5000432461
Encompass_Preferred
Paclitaxel is a naturally occurring antineoplastic agent that stabilizes tubulin polymerization. It can induce mitotic arrest and apoptotic cell death, and also autophagy.
  • Stabilizes tubulin polymerization
  • Induces mitotic arrest
  • Induces apoptotic cell death
  • Induces autophagy
  • Used in cardiovascular disease research
  • Used in cancer research
Catalog No. 50-004-32461 Supplier Medchemexpress LLC Supplier No. HYB00151G
Paclitaxel is a naturally occurring antineoplastic agent that stabilizes tubulin polymerization. It can induce mitotic arrest and apoptotic cell death, and also autophagy.
  • Stabilizes tubulin polymerization
  • Induces mitotic arrest
  • Induces apoptotic cell death
  • Induces autophagy
  • Used in cardiovascular disease research
  • Used in cancer research