Learn More
Medchemexpress LLC Paclitaxel | 33069-62-4 | MFCD00869953 | 100.0% | 853.91 g·mol⁻¹ | C47H51NO14 | 10 MG
SDP
Catalog No. 5000298446
Encompass_Preferred
Paclitaxel is a microtubule-stabilizing antineoplastic reagent used in biological research to inhibit mitosis and induce apoptosis. It is supplied as a solid for preparation of experimental stock solutions and is commonly used in cell culture, in vivo studies, and assay development.
  • Stabilizes tubulin polymerization and induces mitotic arrest.
  • Used for cell culture, in vivo tumor models, and assay development.
  • Supplied as a solid powder suitable for dissolution in DMSO or other organic solvents.
  • Storage recommendations: protect from light; solids at 4°C; solutions at -20°C (short term) or -80°C (long term).
  • Analytical identity confirmed by LC-MS; batch-specific purity reported by certificate of analysis.
Catalog No. 50-002-98446 Supplier Medchemexpress LLC Supplier No. HYB001510MG
Paclitaxel is a microtubule-stabilizing antineoplastic reagent used in biological research to inhibit mitosis and induce apoptosis. It is supplied as a solid for preparation of experimental stock solutions and is commonly used in cell culture, in vivo studies, and assay development.
  • Stabilizes tubulin polymerization and induces mitotic arrest.
  • Used for cell culture, in vivo tumor models, and assay development.
  • Supplied as a solid powder suitable for dissolution in DMSO or other organic solvents.
  • Storage recommendations: protect from light; solids at 4°C; solutions at -20°C (short term) or -80°C (long term).
  • Analytical identity confirmed by LC-MS; batch-specific purity reported by certificate of analysis.