Learn More
Medchemexpress LLC Paclitaxel | 33069-62-4 | MFCD00869953 | 99.9% | 853.91 g/mol | C47H51NO14 | 10 MG
SDP
Catalog No. 5000298873
Encompass_Preferred
Paclitaxel (Standard) is an analytical reference standard of paclitaxel for research and analytical applications. Paclitaxel is a naturally occurring antineoplastic agent that stabilizes tubulin polymerization and can cause mitotic arrest, apoptotic cell death, and autophagy. Provided in a 10 mg reference pack with high-purity material suitable for method development and quantitative assays.
  • Analytical standard intended for research and analytical applications.
  • High purity: 99.88%.
  • Suitable for HPLC and LC-MS method development and validation.
  • Mechanism: stabilizes tubulin polymerization and induces apoptosis.
  • Molecular formula C47H51NO14 and molecular weight 853.91 g/mol.
Catalog No. 50-002-98873 Supplier Medchemexpress LLC Supplier No. HYB0015R10MG
Paclitaxel (Standard) is an analytical reference standard of paclitaxel for research and analytical applications. Paclitaxel is a naturally occurring antineoplastic agent that stabilizes tubulin polymerization and can cause mitotic arrest, apoptotic cell death, and autophagy. Provided in a 10 mg reference pack with high-purity material suitable for method development and quantitative assays.
  • Analytical standard intended for research and analytical applications.
  • High purity: 99.88%.
  • Suitable for HPLC and LC-MS method development and validation.
  • Mechanism: stabilizes tubulin polymerization and induces apoptosis.
  • Molecular formula C47H51NO14 and molecular weight 853.91 g/mol.