Learn More
TARGETMOL CHEMICALS INC Paclitaxel 500MG
SDP
Catalog No. 502880527
Encompass_Preferred
Also available in 1 g, 1 mL, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Paclitaxel (Taxol) is a natural product and a microtubule polymer stabilizer. Paclitaxel has anti-tumor activity and causes cell death by inducing mitotic arrest, apoptosis, and cell autophagy. Purity 99.92%
Catalog No. 50-288-0527 Supplier TARGETMOL CHEMICALS INC Supplier No. T0968500MG
Also available in 1 g, 1 mL, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Paclitaxel (Taxol) is a natural product and a microtubule polymer stabilizer. Paclitaxel has anti-tumor activity and causes cell death by inducing mitotic arrest, apoptosis, and cell autophagy. Purity 99.92%