Learn More
Cayman Chemical Paclitaxel, 33069-62-4, 5 mg
SDP
Supplier:  Cayman Chemical 104615
Encompass_Preferred
Molecular Formula C47H51NO14, Purity ≥98%, Formula Weight 853.9, Synonyms: NSC 125973; A potent disruptor of microtubules used as a chemotherapeutic compound; inhibits growth of cervical (HeLa), lung (A549), breast (MCF-7), colon (HT-29), ovarian (OVG-1), and pancreatic (PC-Sh) carcinomas with IC50 values ranging from 2.5-7.5 nM; induces mitotic arrest and initiates apoptosis of cancer cells through multiple mechanisms.
Catalog No. NC1065970