Learn More
AdipoGen Paclitaxel
SDP
Catalog No. 502560203
Encompass_Preferred
Chemical. CAS 33069-62-4. Formula C47H51NO14. MW 853.91. Synthetic. Anticancer compound. Chemotherapeutic used in patients with cancer and advanced forms of Kaposis sarcoma. Microtubule assembly stabilizer. Reversibly binds to polymerized tubulin. Mitotic spindle assembly, chromosome segregation and cell division inhibitor. Induces cell cycle arrest at the G2/M phase. Apoptosis inducer through aberrant activation of cyclin-dependent kinases CKDs and the c-Jun N-terminal kinase/stress activated protein kinase JNK/SAPK. Immunosuppressor, immunostimulant and TRAIL sensitizer.
Catalog No. 50-256-0203 Supplier AdipoGen Supplier No. CDXP0188M050
Chemical. CAS 33069-62-4. Formula C47H51NO14. MW 853.91. Synthetic. Anticancer compound. Chemotherapeutic used in patients with cancer and advanced forms of Kaposis sarcoma. Microtubule assembly stabilizer. Reversibly binds to polymerized tubulin. Mitotic spindle assembly, chromosome segregation and cell division inhibitor. Induces cell cycle arrest at the G2/M phase. Apoptosis inducer through aberrant activation of cyclin-dependent kinases CKDs and the c-Jun N-terminal kinase/stress activated protein kinase JNK/SAPK. Immunosuppressor, immunostimulant and TRAIL sensitizer.