Learn More
Selleck Chemical LLC Paclitaxel S1150-1g
SDP
Catalog No. 503108209
Encompass_Preferred
Paclitaxel is a microtubule polymer stabilizer with IC50 of 0 1 pM in human endothelial cells Paclitaxel can cause both mitotic arrest and apoptotic cell death Paclitaxel also induces autophagy
Catalog No. 50-310-8209 Supplier Selleck Chemical LLC Supplier No. S11501G
Paclitaxel is a microtubule polymer stabilizer with IC50 of 0 1 pM in human endothelial cells Paclitaxel can cause both mitotic arrest and apoptotic cell death Paclitaxel also induces autophagy