Learn More
Selleck Chemical LLC Paclitaxel S1150-100mg
SDP
Catalog No. 503108494
Encompass_Preferred
Paclitaxel is a microtubule polymer stabilizer with IC50 of 0 1 pM in human endothelial cells Paclitaxel can cause both mitotic arrest and apoptotic cell death Paclitaxel also induces autophagy
Catalog No. 50-310-8494 Supplier Selleck Chemical LLC Supplier No. S1150100MG
Paclitaxel is a microtubule polymer stabilizer with IC50 of 0 1 pM in human endothelial cells Paclitaxel can cause both mitotic arrest and apoptotic cell death Paclitaxel also induces autophagy