Learn More
Abcam Paclita x el, Anticancer agent, 10MG
Catalog No. 502584678
Encompass_Preferred
MW 853.9 Da, Purity >98%. Anticancer agent. Inhibits depolymerization of microtubules, causing mitotic arrest in cancer cells, and apoptosis.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-4678 Supplier Abcam Supplier No. AB12014310MG
MW 853.9 Da, Purity >98%. Anticancer agent. Inhibits depolymerization of microtubules, causing mitotic arrest in cancer cells, and apoptosis.
The product is subject to the following: Abcam Restricted Use Statement