Learn More
Abcam Paclita x el, Anticancer agent, 50MG
Catalog No. 502584677
Encompass_Preferred
MW 853.9 Da, Purity >98%. Anticancer agent. Inhibits depolymerization of microtubules, causing mitotic arrest in cancer cells, and apoptosis.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-4677 Supplier Abcam Supplier No. AB12014350MG
MW 853.9 Da, Purity >98%. Anticancer agent. Inhibits depolymerization of microtubules, causing mitotic arrest in cancer cells, and apoptosis.
The product is subject to the following: Abcam Restricted Use Statement