Learn More
Medchemexpress LLC Paclitaxel-d5 | 1129540-33-5 | 1129540-33-5 | 99.9% | C47H46D5NO14 | 1 MG
SDP
Catalog No. 5000432801
Encompass_Preferred
Paclitaxel-d5 is a deuterium-labeled Paclitaxel, a natural antineoplastic agent that works by stabilizing tubulin polymerization. This product is strictly for research purposes and is not intended for patient use. Deuteration, the process of incorporating stable heavy isotopes into drug molecules, is primarily employed as a tracer for quantitation during the drug development process and can impact the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Stabilizes tubulin polymerization
  • For research use only
Catalog No. 50-004-32801 Supplier Medchemexpress LLC Supplier No. HYB0015S1MG
Paclitaxel-d5 is a deuterium-labeled Paclitaxel, a natural antineoplastic agent that works by stabilizing tubulin polymerization. This product is strictly for research purposes and is not intended for patient use. Deuteration, the process of incorporating stable heavy isotopes into drug molecules, is primarily employed as a tracer for quantitation during the drug development process and can impact the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Stabilizes tubulin polymerization
  • For research use only