Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Paclitaxel-d5 | 1129540-33-5 | 98.1% | 858.94 | C47H46D5NO14 | 10 MG
SDP

Catalog No. 5000298874
Encompass_Preferred

Paclitaxel-d5 is a deuterium-labeled analog of paclitaxel supplied as a white to off-white solid for use as a tracer and internal standard in quantitative analytical methods. It stabilizes tubulin polymerization and is intended for research use in NMR, GC-MS, and LC-MS applications.

  • High purity (98.1%).
  • Molecular weight 858.94 Da.
  • White to off-white solid form.
  • Soluble in DMSO and DMF; moderately soluble in ethanol.
  • Stable storage as powder at -20 °C for long-term retention.
  • Designed for use as tracer and internal standard in quantitative assays.

Catalog No. 50-002-98874 Supplier Medchemexpress LLC Supplier No. HYB0015S10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Paclitaxel-d5 is a deuterium-labeled analog of paclitaxel supplied as a white to off-white solid for use as a tracer and internal standard in quantitative analytical methods. It stabilizes tubulin polymerization and is intended for research use in NMR, GC-MS, and LC-MS applications.

  • High purity (98.1%).
  • Molecular weight 858.94 Da.
  • White to off-white solid form.
  • Soluble in DMSO and DMF; moderately soluble in ethanol.
  • Stable storage as powder at -20 °C for long-term retention.
  • Designed for use as tracer and internal standard in quantitative assays.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.