Learn More
Medchemexpress LLC Paclitaxel-d5 | 1129540-33-5 | 99.9% | C47H46D5NO14 | 5 MG
SDP
Catalog No. 5000432160
Encompass_Preferred
Paclitaxel-d5 is a deuterium-labeled Paclitaxel, a naturally occurring antineoplastic agent that stabilizes tubulin polymerization. This compound is suitable for research applications.
  • Can be used as a tracer
  • Serves as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuteration can impact pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-32160 Supplier Medchemexpress LLC Supplier No. HYB0015S5MG
Paclitaxel-d5 is a deuterium-labeled Paclitaxel, a naturally occurring antineoplastic agent that stabilizes tubulin polymerization. This compound is suitable for research applications.
  • Can be used as a tracer
  • Serves as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuteration can impact pharmacokinetic and metabolic profiles of drugs