Learn More
Medchemexpress LLC Paclitaxel-d5 | 1129540-33-5 | 99.9% | C47H46D5NO14 | 500 UG
SDP
Catalog No. 5000432027
Encompass_Preferred
Paclitaxel-d5 is a deuterium-labeled Paclitaxel, a naturally occurring antineoplastic agent that stabilizes tubulin polymerization. Deuteration has garnered attention due to its potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Incorporates stable heavy isotopes of hydrogen, carbon, and other elements into drug molecules as tracers for quantitation during the drug development process.
Catalog No. 50-004-32027 Supplier Medchemexpress LLC Supplier No. HYB0015S500UG
Paclitaxel-d5 is a deuterium-labeled Paclitaxel, a naturally occurring antineoplastic agent that stabilizes tubulin polymerization. Deuteration has garnered attention due to its potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Incorporates stable heavy isotopes of hydrogen, carbon, and other elements into drug molecules as tracers for quantitation during the drug development process.