Learn More
Medchemexpress LLC Paclitaxel-d5 (benzoyloxy) | 1261254-56-1 | 98.1% | C47H46D5NO14 | 500 UG
SDP
Catalog No. 5000432821
Encompass_Preferred
Paclitaxel-d5 (benzoyloxy) is the deuterium-labeled Paclitaxel. Paclitaxel is a naturally occurring antineoplastic agent that stabilizes tubulin polymerization. It can cause both mitotic arrest and apoptotic cell death and also induces autophagy. This product is for research use only and not sold to patients.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-32821 Supplier Medchemexpress LLC Supplier No. HYB0015S1500UG
Paclitaxel-d5 (benzoyloxy) is the deuterium-labeled Paclitaxel. Paclitaxel is a naturally occurring antineoplastic agent that stabilizes tubulin polymerization. It can cause both mitotic arrest and apoptotic cell death and also induces autophagy. This product is for research use only and not sold to patients.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.