Learn More
Hanna Pharmaceutical Supply Co., Inc. PACLITAXEL IV ST NON-VNTD 48CS
Supplier:  Hanna Pharmaceutical Supply Co., Inc. 2C8858
Encompass_Preferred
Paclitaxel IV Set Non-vented 1 Port 10 Drops / mL Drip Rate 0.22 Micron Filter 107 Inch Tubing Solution
Catalog No. NC3103979