Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Paclitaxel MP Biomedicals

Catalog No. ICN19353280 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 mg
5 mg
25 mg
100 mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19353280 100 mg
ICN19353201 1 mg
ICN19353205 5 mg
ICN19353225 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. ICN19353280 Supplier MP Biomedicals, Inc Supplier No. 0219353280
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 373-49-9
Molecular Formula C16H30O2
Molecular Weight (g/mol) 254.41
MDL Number MFCD00004437
InChI Key SECPZKHBENQXJG-FPLPWBNLSA-N
Synonym palmitoleic acid, cis-9-hexadecenoic acid, z-hexadec-9-enoic acid, 9-cis-hexadecenoic acid, zoomaric acid, z-9-hexadecenoic acid, 9z-hexadecenoic acid, palmitolinoleic acid, cis-palmitoleic acid, zoomeric acid
PubChem CID 445638
ChEBI CHEBI:28716
SMILES CCCCCC\C=C/CCCCCCCC(O)=O

Specifications

Color White
Assay Percent Range ≥98%
Quantity 100 mg
IUPAC Name (Z)-hexadec-9-enoic acid
Formula Weight 853.9
Percent Purity ≥98%
Physical Form Powder
Chemical Name or Material Paclitaxel, From the Pacific Yew Tree
EINECSNumber 200-882-9
Recommended Storage Store at room temperature (15° to 30°C)
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.