Learn More
Cayman Chemical Paclitaxel, Purity: ≥98%, CAS: 33069-62-4, 25 mg
SDP
Catalog No. 502887821
Encompass_Preferred
A potent disruptor of microtubules used as a chemotherapeutic compound; inhibits growth of cervical (HeLa), lung (A549), breast (MCF-7), colon (HT-29), ovarian (OVG-1), and pancreatic (PC-Sh) carcinomas with IC50 values ranging from 2.5-7.5 nM; induces mitotic arrest and initiates apoptosis of cancer cells through multiple mechanisms
Catalog No. 50-288-7821 Supplier Cayman Chemical Supplier No. 1046125 MG
A potent disruptor of microtubules used as a chemotherapeutic compound; inhibits growth of cervical (HeLa), lung (A549), breast (MCF-7), colon (HT-29), ovarian (OVG-1), and pancreatic (PC-Sh) carcinomas with IC50 values ranging from 2.5-7.5 nM; induces mitotic arrest and initiates apoptosis of cancer cells through multiple mechanisms