Learn More
Cayman Chemical Paclitaxel, Purity: ≥98%, CAS: 33069-62-4, 100 mg
SDP
Catalog No. 502889648
Encompass_Preferred
A potent disruptor of microtubules used as a chemotherapeutic compound; inhibits growth of cervical (HeLa), lung (A549), breast (MCF-7), colon (HT-29), ovarian (OVG-1), and pancreatic (PC-Sh) carcinomas with IC50 values ranging from 2.5-7.5 nM; induces mitotic arrest and initiates apoptosis of cancer cells through multiple mechanisms
Catalog No. 50-288-9648 Supplier Cayman Chemical Supplier No. 10461100 MG
A potent disruptor of microtubules used as a chemotherapeutic compound; inhibits growth of cervical (HeLa), lung (A549), breast (MCF-7), colon (HT-29), ovarian (OVG-1), and pancreatic (PC-Sh) carcinomas with IC50 values ranging from 2.5-7.5 nM; induces mitotic arrest and initiates apoptosis of cancer cells through multiple mechanisms