Learn More
Medchemexpress LLC Paclitaxel (Standard) | 33069-62-4 | 99.9% | C₄₇H₅₁NO₁₄ | 25 MG
SDP
Catalog No. 5000359070
Encompass_Preferred
Paclitaxel (Standard) is an analytical standard of Paclitaxel, intended for research and analytical applications. It is a naturally occurring antineoplastic agent that stabilizes tubulin polymerization, causing mitotic arrest, apoptotic cell death, and inducing autophagy.
  • Used as an analytical standard for qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
  • Naturally occurring antineoplastic agent.
  • Stabilizes tubulin polymerization.
  • Causes mitotic arrest and apoptotic cell death.
  • Induces autophagy.
Catalog No. 50-003-59070 Supplier Medchemexpress LLC Supplier No. 25MGHYB0015R
Paclitaxel (Standard) is an analytical standard of Paclitaxel, intended for research and analytical applications. It is a naturally occurring antineoplastic agent that stabilizes tubulin polymerization, causing mitotic arrest, apoptotic cell death, and inducing autophagy.
  • Used as an analytical standard for qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
  • Naturally occurring antineoplastic agent.
  • Stabilizes tubulin polymerization.
  • Causes mitotic arrest and apoptotic cell death.
  • Induces autophagy.