Learn More
Medchemexpress LLC Paclitaxel (Standard) | 33069-62-4 | 99.9% | C₄₇H₅₁NO₁₄ | 5 MG
SDP
Catalog No. 5000432045
Encompass_Preferred
Paclitaxel (Standard) is an analytical standard of Paclitaxel, a naturally occurring antineoplastic agent that stabilizes tubulin polymerization, causing mitotic arrest and apoptotic cell death, and also induces autophagy. This compound serves as a reference standard for various research applications.
  • Analytical standard for research and analytical applications
  • Stabilizes tubulin polymerization
  • Induces mitotic arrest and apoptotic cell death
  • Induces autophagy
  • Reference standard for assays
  • Used in HPLC, GC, and MS experiments
  • White to off-white solid appearance
  • Molecular weight is 853.91
  • Derived from Taxaceae family plants (*Taxus chinensis*)
Catalog No. 50-004-32045 Supplier Medchemexpress LLC Supplier No. HYB0015R5MG
Paclitaxel (Standard) is an analytical standard of Paclitaxel, a naturally occurring antineoplastic agent that stabilizes tubulin polymerization, causing mitotic arrest and apoptotic cell death, and also induces autophagy. This compound serves as a reference standard for various research applications.
  • Analytical standard for research and analytical applications
  • Stabilizes tubulin polymerization
  • Induces mitotic arrest and apoptotic cell death
  • Induces autophagy
  • Reference standard for assays
  • Used in HPLC, GC, and MS experiments
  • White to off-white solid appearance
  • Molecular weight is 853.91
  • Derived from Taxaceae family plants (*Taxus chinensis*)